Gold trichloride 99%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Gold(III) chloride, Gold (III) chloride
318645000 318640050
ACRO318645000EA 170 EUR
ACRO318645000 ACRO318640050
Gold trichloride 99%
Gold trichloride
Formula: AuCl₃
MW: 303.32 g/mol
Melting Pt: 254 °C
Density: 4.3
Storage Temperature: Ambient
MDL Number: MFCD00014172
CAS Number: 13453-07-1
EINECS: 236-623-1
Merck Index: 13,04536
Order Now
Specification Test Results
Appearance Orange-red to brown to violet Crystalline powder
Assay ≥98.5 % (ex Au)