Gold trichloride, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Gold(III) chloride, Gold (III) chloride
M-Clarity™ quality level = MQ100
379948-1G
379948-250MG
SIAL379948-1GEA
215
EUR
SIAL379948-1G
SIAL379948-250MG
Gold trichloride, Sigma-Aldrich®
Gold trichloride
Formula:
AuCl₃ MW: 303.32 g/mol Melting Pt: 287…288 °C Density: 4.3 g/cm³ (20 °C) |
MDL Number:
MFCD00014172 CAS Number: 13453-07-1 EINECS: 236-623-1 UN: 3260 ADR: 8,III Merck Index: 13,04536 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...