Copper(II) chloride, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Copper dichloride, Cupric chloride
M-Clarity™ quality level = MQ100
203149-10G 203149-50G
SIAL203149-10GEA 225 EUR
SIAL203149-10G SIAL203149-50G
Copper(II) chloride, Sigma-Aldrich®
Copper (II) chloride
Formula: Cl₂Cu
Boiling Pt: 1365.85 °C (1013 hPa)
Melting Pt: 628 °C
Density: 3.054 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010972
CAS Number: 7447-39-4
EINECS: 231-210-2
UN: 2802
ADR: 8,III
Merck Index: 13,02660