Copper(II) chloride, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
222011-1KG 222011-250G 222011-50G
SIAL222011-1KGEA 189 EUR
SIAL222011-1KG SIAL222011-250G SIAL222011-50G
Copper(II) chloride, Sigma-Aldrich®
Formula: Cl₂Cu
Boiling Pt: 1365.85 °C (1013 hPa)
Melting Pt: 628 °C
Density: 3.054 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010972
CAS Number: 7447-39-4
EINECS: 231-210-2
UN: 2802
ADR: 8,III
Merck Index: 13,02660