Silver chromate, Sigma-Aldrich®
Supplier: Merck
227897-10G 227897-50G
SIAL227897-10GEA 97.2 EUR
SIAL227897-10G SIAL227897-50G
Silver chromate, Sigma-Aldrich®
Silver chromate
Formula: Ag₂CrO₄
Boiling Pt: 1550 °C (1013 hPa)
Density: 5.536 g/cm³ (25 °C)
MDL Number: MFCD00003402
CAS Number: 7784-01-2
EINECS: 232-043-8
UN: 1479
ADR: 5.1,III