Silver fluoride, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Silver monofluoride, Silver(I) fluoride, Argentous fluoride
M-Clarity™ quality level = MQ100
226858-1G 226858-25G 226858-5G
SIAL226858-1GEA 43 EUR
SIAL226858-1G SIAL226858-25G SIAL226858-5G
Silver fluoride, Sigma-Aldrich®
Silver fluoride
Formula: AgF
Boiling Pt: 1150 °C (1013 hPa)
Melting Pt: 433 °C
Density: 5.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003410
CAS Number: 7775-41-9
EINECS: 231-895-8
UN: 3260
ADR: 8,II
Merck Index: 12,08657