Copper(I) iodide, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Cuprous iodide
M-Clarity™ quality level = MQ200
215554-100G
215554-25G
215554-5G
SIAL215554-100GEA
433
EUR
SIAL215554-100G
SIAL215554-25G
SIAL215554-5G
Copper(I) iodide, Sigma-Aldrich®
Copper (I) iodide
Formula:
CuI MW: 190.45 g/mol Boiling Pt: 1290 Melting Pt: 602 Density: 5.62 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010978 CAS Number: 7681-65-4 EINECS: 231-674-6 UN: 3077 ADR: 9 III Merck Index: 13,02692 |