Copper(II) oxide

Supplier: Roth Carl

ROTH4467.1EA 86.1 EUR
ROTH4467.1
Copper(II) oxide
Copper(II) oxide
Formula: CuO
MW: 79,55 g/mol
Boiling Pt: 2000 °C (1013 hPa)
Melting Pt: 1326 °C
Density: 4,9 g/cm³ (20 °C)
MDL Number: MFCD00010979
UN: 3077
ADR: 9,III
Merck Index: 13,02674

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR