Copper(II) oxide
Supplier: Roth Carl
Warning
8169.1 8169.2
ROTH8169.1EA 72 EUR
ROTH8169.1 ROTH8169.2
Copper(II) oxide
Copper (II) oxide
Formula: CuO
Boiling Pt: 2000 °C (1013 hPa)
Melting Pt: 1326 °C
Density: 4.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010979
CAS Number: 1317-38-0
EINECS: 215-269-1
UN: 3077
ADR: 9,III
Merck Index: 13,02674