Copper(II) oxide, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Cupric oxide, Copper oxide, Copper monoxide granulated
M-Clarity™ quality level = MQ100
203130-25G 203130-5G
SIAL203130-25GEA 345 EUR
SIAL203130-25G SIAL203130-5G
Copper(II) oxide, Sigma-Aldrich®
Copper (II) oxide
Formula: CuO
MW: 79.55 g/mol
Boiling Pt: 2000
Melting Pt: 1326
Density: 4.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010979
CAS Number: 1317-38-0
EINECS: 215-269-1
UN: 3077
ADR: 9 III
Merck Index: 13,02674