Copper(II) oxide ≥98%, powder <10 μm, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

SIAL208841-12KGEA 1120 EUR
SIAL208841-12KG
Copper(II) oxide ≥98%, powder <10 μm, Sigma-Aldrich®
Copper(II) oxide
Formula: CuO
MW: 79,55 g/mol
Boiling Pt: 2000 °C (1013 hPa)
Melting Pt: 1326 °C
Density: 4,9 g/cm³ (20 °C)
MDL Number: MFCD00010979
UN: 3077
ADR: 9,III
Merck Index: 13,02674

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR