Silver(I) sulphide, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
241474-25G 241474-5G
SIAL241474-25GEA 381 EUR
SIAL241474-25G SIAL241474-5G
Silver(I) sulphide, Sigma-Aldrich®
Silver (I) sulphide
Formula: Ag₂S
Melting Pt: 842 °C
Density: 7.23 g/cm³ (25 °C)
MDL Number: MFCD00003406
CAS Number: 21548-73-2
EINECS: 244-438-2
Merck Index: 12,08677