Mercury(II) chloride
Supplier: Roth Carl
Danger
Synonyms: Mercuric chloride, Mercury Chloride, Mercury dichloride
7904.1 7904.2
ROTH7904.1EA 118 EUR
ROTH7904.1 ROTH7904.2
Mercury(II) chloride
Mercury (II) chloride
Formula: HgCl₂
MW: 271.5 g/mol
Boiling Pt: 302 °C (1013 hPa)
Melting Pt: 273 °C
Density: 5.44 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011041
CAS Number: 7487-94-7
EINECS: 231-299-8
UN: 1624
ADR: 6.1,II
Merck Index: 13,05901