Zinc chloride ≥98% Ph. Eur., purified
Supplier: VWR Chemicals
29144.365EA
0
EUR
29144.365
Zinc chloride ≥98% Ph. Eur., purified
Zinc chloride
Formula:
ZnCl₂ MW: 136,3 g/mol Boiling Pt: 732 °C (1013 hPa) Melting Pt: 283 °C Density: 2,91 g/cm³ (20 °C) |
MDL Number:
MFCD00011295 UN: 2331 ADR: 8,III Merck Index: 13,10185 |
Specification Test Results
Assay | Min. 98 % |
Fe (Iron) | Max. 20 ppm |
Pb (Lead) | Max. 20 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...