Cadmium(II) carbonate, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
289140-100G 289140-500G
SIAL289140-100GEA 57.2 EUR
SIAL289140-100G SIAL289140-500G
Cadmium(II) carbonate, Sigma-Aldrich®
Cadmium (II) carbonate
Formula: CCdO₃
Melting Pt: 357 °C
Density: 5.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010918
CAS Number: 513-78-0
EINECS: 208-168-9
UN: 2570
ADR: 6.1,III
Merck Index: 12,01652