Cadmium(II) iodide 99%
Supplier: Thermo Fisher Scientific
|
Danger
|
211462500
211460010
ACRO211462500EA
156
EUR
ACRO211462500
ACRO211460010
Cadmium(II) iodide 99%
Cadmium (II) iodide
Formula:
CdI₂ Boiling Pt: 787 °C (760 mmHg) Melting Pt: 388 °C Density: 5,67 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010920 CAS Number: 7790-80-9 EINECS: 232-223-6 UN: 2570 ADR: 6.1,III Merck Index: 12,01657 |
Specification Test Results
Appearance | White to light yellow Powder or crystals and/or flakes |
Titration Complexometric | ≥98.5 % |