Mercury(II) iodide, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

SIAL203785-25GEA 290 EUR
SIAL203785-25G
Mercury(II) iodide, Sigma-Aldrich®
Mercury(II) iodide
Formula: HgI₂
MW: 454,4 g/mol
Boiling Pt: 354 °C (1013 hPa)
Melting Pt: 259 °C
Density: 6,36 g/cm³ (20 °C)
MDL Number: MFCD00011044
CAS Number: 7774-29-0
ADR: 6.1,II
Merck Index: 12,05932

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR