Zinc iodide, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Zinc (II) iodide
MQ100
466360-25G 466360-5G
SIAL466360-25GEA 534 EUR
SIAL466360-25G SIAL466360-5G
Zinc iodide, Sigma-Aldrich®
Zinc iodide
Formula: I₂Zn
MW: 319.2 g/mol
Boiling Pt: 620
Melting Pt: 446
Density: 4.74 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011299
CAS Number: 10139-47-6
EINECS: 233-396-0
UN: 3260
ADR: 8 II
Merck Index: 12,10269