Zinc iodide, Sigma-Aldrich®

Supplier: Merck
Warning

96483-100G 96483-500G
SIAL96483-100GEA 108 EUR
SIAL96483-100G SIAL96483-500G
Zinc iodide, Sigma-Aldrich®
Zinc iodide
Formula: I₂Zn
Boiling Pt: 620 °C (1013 hPa)
Melting Pt: 446 °C
Density: 4.74 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011299
CAS Number: 10139-47-6
EINECS: 233-396-0
UN: 3260
ADR: 8,II
Merck Index: 12,10269

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR