Cadmium oxide, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Cadmium (II) oxide
M-Clarity™ quality level = MQ100
202894-25G 202894-5G
SIAL202894-25GEA 168 EUR
SIAL202894-25G SIAL202894-5G
Cadmium oxide, Sigma-Aldrich®
Cadmium oxide
Formula: CdO
MW: 128.41 g/mol
Boiling Pt: 1559
Melting Pt: ∼1426 °C
Density: 8.15 g/cm³ (20 °C)
MDL Number: MFCD00010921
CAS Number: 1306-19-0
EINECS: 215-146-2
UN: 2570
ADR: 6.1 I
Merck Index: 13,01623