Cadmium oxide, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Cadmium (II) oxide
M-Clarity™ quality level = MQ200
244783-100G 244783-500G
SIAL244783-100GEA 72.3 EUR
SIAL244783-100G SIAL244783-500G
Cadmium oxide, Sigma-Aldrich®
Cadmium oxide
Formula: CdO
MW: 128.41 g/mol
Boiling Pt: 1559
Melting Pt: ∼1426 °C
Density: 8.15 g/cm³ (20 °C)
MDL Number: MFCD00010921
CAS Number: 1306-19-0
EINECS: 215-146-2
UN: 2570
ADR: 6.1 I
Merck Index: 13,01623