Zinc sulphide, Sigma-Aldrich®
Supplier: Merck
Synonyms: Zinc (II) sulphide
M-Clarity™ quality level = MQ100
14459-1KG 14459-500G
SIAL14459-1KGEA 341 EUR
SIAL14459-1KG SIAL14459-500G
Zinc sulphide, Sigma-Aldrich®
Zinc sulphide
Formula: SZn
MW: 97.46 g/mol
Melting Pt: 1064
Density: 4.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011301
CAS Number: 1314-98-3
EINECS: 215-251-3
Merck Index: 12,10294