Mercury(II) thiocyanate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Mercury dithiocyanate, Mercuric thiocyanate, Mercury (II) rhodanide
M-Clarity™ quality level = MQ100
83374-100G 83374-500G 83374-5G
SIAL83374-100GEA 764 EUR
SIAL83374-100G SIAL83374-500G SIAL83374-5G
Mercury(II) thiocyanate, Sigma-Aldrich®
Mercury (II) thiocyanate
Formula: C₂HgN₂S₂
MW: 316.76 g/mol
Melting Pt: –38.9 °C
Density: 4 g/cm³ (20 °C)
Flash Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00011039
CAS Number: 592-85-8
EINECS: 209-773-0
UN: 1646
ADR: 6.1,II
Merck Index: 13,05917