Mercury(II) sulphate
Supplier: Roth Carl
ROTHX871.1EA 129 EUR
ROTHX871.1
Mercury(II) sulphate
Mercury(II) sulphate
Formula: HgSO₄
MW: 296,65 g/mol
Melting Pt: 850 °C
Density: 6,47 g/cm³ (25 °C)
MDL Number: MFCD00011047
CAS Number: 7783-35-9
UN: 1645
ADR: 6.1,II
Merck Index: 13,05914