Scandium(III) chloride, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
409359-1G
409359-5G
SIAL409359-1GEA
268
EUR
SIAL409359-1G
SIAL409359-5G
Scandium(III) chloride, Sigma-Aldrich®
Scandium (III) chloride
Formula:
Cl₃Sc MW: 151.31 g/mol Melting Pt: 940 °C Density: 1.67 g/cm³ (940 °C) |
MDL Number:
MFCD00011221 CAS Number: 10361-84-9 EINECS: 233-799-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...