Discandium trioxide, Sigma-Aldrich®
Supplier: Merck
Synonyms: Scandium oxide, Scandium (III) oxide
M-Clarity™ quality level = MQ100
294020-500MG 294020-5G
SIAL294020-500MGEA 87.6 EUR
SIAL294020-500MG SIAL294020-5G
Discandium trioxide, Sigma-Aldrich®
Discandium trioxide
Formula: O₃Sc₂
Melting Pt: 1000 °C
Density: 3.86 g/cm³ (20 °C)
MDL Number: MFCD00011223
CAS Number: 12060-08-1
EINECS: 235-042-0
Merck Index: 14,08392