Diyttrium trioxide, Sigma-Aldrich®
Supplier: Merck
Synonyms: Yttrium oxide, Yttrium (III) oxide
M-Clarity™ quality level = MQ100
204927-50G 204927-10G
SIAL204927-50GEA 414 EUR
SIAL204927-50G SIAL204927-10G
Diyttrium trioxide, Sigma-Aldrich®
Diyttrium trioxide
Formula: Y₂O₃
MW: 225.81 g/mol
Boiling Pt: 4300 °C (1013 hPa)
Melting Pt: 2410 °C
Density: 5.01 g/cm³ (20 °C)
MDL Number: MFCD00011473
CAS Number: 1314-36-9
EINECS: 215-233-5