Diyttrium trioxide, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
205168-10G 205168-250G 205168-50G
SIAL205168-10GEA 47.2 EUR
SIAL205168-10G SIAL205168-250G SIAL205168-50G
Diyttrium trioxide, Sigma-Aldrich®
Diyttrium trioxide
Formula: Y₂O₃
Boiling Pt: 4300 °C (1013 hPa)
Melting Pt: 2410 °C
Density: 5.01 g/cm³ (20 °C)
MDL Number: MFCD00011473
CAS Number: 1314-36-9
EINECS: 215-233-5