Titanium(IV) oxide, Sigma-Aldrich®
Supplier: Merck
Synonyms: Titanic anhydride, Pigment White 6, Rutile, Titanium(IV) oxide, rutile, Titanium dioxide
M-Clarity™ quality level = MQ100
204730-25G 204730-5G
SIAL204730-25GEA 284 EUR
SIAL204730-25G SIAL204730-5G
Titanium(IV) oxide, Sigma-Aldrich®
Titanium (IV) oxide
Formula: TiO₂
MW: 79.88 g/mol
Boiling Pt: 2900 °C (1013 hPa)
Melting Pt: 1855 °C
Density: 4.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011269
CAS Number: 13463-67-7
EINECS: 236-675-5
Merck Index: 13,09549