Titanium(IV) butanolate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Titanium (IV) butoxide, Titanium tetrabutanolate, Tetra-n-butyl orthotitanate, Titanium (IV) n-butoxide
M-Clarity™ quality level = MQ200
244112-100G 244112-2KG 244112-500G 244112-5G
SIAL244112-100GEA 41.7 EUR
SIAL244112-100G SIAL244112-2KG SIAL244112-500G SIAL244112-5G
Titanium(IV) butanolate, Sigma-Aldrich®
Titanium(IV) butanolate
Formula: C₁₆H₃₆O₄Ti
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: –55 °C
Density: 0.993 g/cm³ (20 °C)
Flash Pt: 47 °C
MDL Number: MFCD00009433
CAS Number: 5593-70-4
EINECS: 227-006-8
UN: 1993
ADR: 3,III