Titanium(IV) butanolate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Titanium (IV) butoxide, Titanium tetrabutanolate, Tetra-n-butyl orthotitanate, Titanium (IV) n-butoxide
M-Clarity™ quality level = MQ200
86910-1L 86910-250ML
SIAL86910-1LEA 172 EUR
SIAL86910-1L SIAL86910-250ML
Titanium(IV) butanolate, Sigma-Aldrich®
Titanium(IV) butanolate
Formula: C₁₆H₃₆O₄Ti
MW: 340.34 g/mol
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: –55 °C
Density: 0.993 g/cm³ (20 °C)
Flash Pt: 47 °C
MDL Number: MFCD00009433
CAS Number: 5593-70-4
EINECS: 227-006-8
UN: 1993
ADR: 3,III