Tantalum pentachloride, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Tantalum (V) chloride
M-Clarity™ quality level = MQ200
218634-100G
218634-25G
218634-5G
SIAL218634-100GEA
682
EUR
SIAL218634-100G
SIAL218634-25G
SIAL218634-5G
Tantalum pentachloride, Sigma-Aldrich®
Tantalum pentachloride
Formula:
Cl₅Ta Boiling Pt: 232…235 °C (1013 hPa) Melting Pt: 216…217 °C Density: 3.68 g/cm³ (20 °C) |
MDL Number:
MFCD00011253 CAS Number: 7721-01-9 EINECS: 231-755-6 UN: 3260 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...