Vanadium(III) oxide, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
463744-25G
463744-5G
SIAL463744-25GEA
770
EUR
SIAL463744-25G
SIAL463744-5G
Vanadium(III) oxide, Sigma-Aldrich®
Vanadium (III) oxide
Formula:
O₃V₂ MW: 149.88 g/mol Melting Pt: 1940 °C Density: 4.87 g/cm³ (20 °C) |
MDL Number:
MFCD00011455 CAS Number: 1314-34-7 EINECS: 215-230-9 UN: 3285 ADR: 6.1,II Merck Index: 12,10059 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...