Vanadium(V) oxide, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Vanadium pentoxide, di-Vanadium pentaoxide
M-Clarity™ quality level = MQ100
204854-1G 204854-25G 204854-5G
SIAL204854-1GEA 53.8 EUR
SIAL204854-1G SIAL204854-25G SIAL204854-5G
Vanadium(V) oxide, Sigma-Aldrich®
Vanadium (V) oxide
Formula: O₅V₂
MW: 181.88 g/mol
Boiling Pt: 1750 °C (1013 hPa)
Melting Pt: 690 °C
Density: 3.35 g/cm³ (20 °C)
MDL Number: MFCD00011457
CAS Number: 1314-62-1
EINECS: 215-239-8
UN: 2862
ADR: 6.1,III
Merck Index: 13,09987