Chromium(II) chloride 90%
Supplier: Thermo Scientific
Warning
Synonyms: Chromium dichloride
223810050 223810100
ACRO223810050EA 101 EUR
ACRO223810050 ACRO223810100
Chromium(II) chloride 90%
Chromium (II) chloride
Formula: Cl₂Cr
MW: 122.9 g/mol
Boiling Pt: 1302 °C (1013 hPa)
Melting Pt: 824 °C
Density: 2,87 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010947
CAS Number: 10049-05-5
EINECS: 233-163-3
ADR: 8,III
Merck Index: 13,02264
Order Now
Specification Test Results
Appearance Green to grey Powder
Titration Argentometric ≥88 %