Tungsten(VI) chloride, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Tungsten hexachloride

M-Clarity™ quality level = MQ100

645168-25G 645168-5G
SIAL645168-25GEA 1030 EUR
SIAL645168-25G SIAL645168-5G
Tungsten(VI) chloride, Sigma-Aldrich®
Tungsten (VI) chloride
Formula: Cl₆W
MW: 396.56 g/mol
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 275 °C
Density: 3.518 g/cm³ (20 °C)
MDL Number: MFCD00011463
CAS Number: 13283-01-7
EINECS: 236-293-9
UN: 3260
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR