Molybdenum(IV) sulphide, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Molybdenum disulphide
M-Clarity™ quality level = MQ100
234842-100G
234842-500G
SIAL234842-100GEA
71.4
EUR
SIAL234842-100G
SIAL234842-500G
Molybdenum(IV) sulphide, Sigma-Aldrich®
Molybdenum (IV) sulphide
Formula:
MoS₂ MW: 160.07 g/mol Melting Pt: 2375 °C Density: 4.8 g/cm³ (20 °C) |
MDL Number:
MFCD00003470 CAS Number: 1317-33-5 EINECS: 215-263-9 Merck Index: 12,06318 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...