Molybdenum(IV) sulphide, Sigma-Aldrich®

Supplier: Merck

Synonyms: Molybdenum disulphide

69860-100G 69860-500G
SIAL69860-100GEA 92.6 EUR
SIAL69860-100G SIAL69860-500G
Molybdenum(IV) sulphide, Sigma-Aldrich®
Molybdenum (IV) sulphide
Formula: MoS₂
MW: 160.07 g/mol
Melting Pt: 2375 °C
Density: 4.8 g/cm³ (20 °C)
MDL Number: MFCD00003470
CAS Number: 1317-33-5
EINECS: 215-263-9
Merck Index: 12,06318

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR