Cobalt(II) bromide, Sigma-Aldrich®
Supplier: Merck
Synonyms: Cobaltous bromide
M-Clarity™ quality level = MQ100
334022-250G 334022-50G
SIAL334022-250GEA 323 EUR
SIAL334022-250G SIAL334022-50G
Cobalt(II) bromide, Sigma-Aldrich®
Cobalt (II) bromide
Formula: Br₂Co
MW: 218.74 g/mol
Density: 4.909 g/cm³ (20 °C)
MDL Number: MFCD00016017
CAS Number: 7789-43-7
EINECS: 232-166-7
Merck Index: 12,02496