Cobalt(II) bromide, Sigma-Aldrich®
Supplier: Merck
Synonyms: Cobaltous bromide
M-Clarity™ quality level = MQ100
427136-1G 427136-5G
SIAL427136-1GEA 101 EUR
SIAL427136-1G SIAL427136-5G
Cobalt(II) bromide, Sigma-Aldrich®
Cobalt (II) bromide
Formula: Br₂Co
MW: 218.74 g/mol
Density: 4.909 g/cm³ (20 °C)
MDL Number: MFCD00016017
CAS Number: 7789-43-7
EINECS: 232-166-7
Merck Index: 12,02496