Iron(II) chloride, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
450936-25G 450936-5G
SIAL450936-25GEA 331 EUR
SIAL450936-25G SIAL450936-5G
Iron(II) chloride, Sigma-Aldrich®
Iron (II) chloride
Formula: Cl₂Fe
Boiling Pt: 2023 °C (1013 hPa)
Melting Pt: 674 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011004
CAS Number: 7758-94-3
EINECS: 231-843-4
UN: 3260
ADR: 8,III
Merck Index: 12,04091