Iron(II) chloride, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
450944-10G 450944-50G
SIAL450944-10GEA 160 EUR
SIAL450944-10G SIAL450944-50G
Iron(II) chloride, Sigma-Aldrich®
Iron (II) chloride
Formula: Cl₂Fe
Boiling Pt: 2023 °C (1013 hPa)
Melting Pt: 674 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011004
CAS Number: 7758-94-3
EINECS: 231-843-4
UN: 3260
ADR: 8,III
Merck Index: 12,04091