Palladium(II) chloride, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Palladium dichloride
MQ200
205885-25G 205885-1G 205885-5G
SIAL205885-25GEA 2140 EUR
SIAL205885-25G SIAL205885-1G SIAL205885-5G
Palladium(II) chloride, Sigma-Aldrich®
Palladium (II) chloride
Formula: PdCl₂
MW: 177.33 g/mol
Melting Pt: 678
Density: 4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003558
CAS Number: 7647-10-1
EINECS: 231-596-2
UN: 3260
ADR: 8 III
Merck Index: 13,07058