Palladium(II) chloride, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Palladium dichloride
MQ100
520659-1G 520659-25G 520659-5G
SIAL520659-1GEA 146 EUR
SIAL520659-1G SIAL520659-25G SIAL520659-5G
Palladium(II) chloride, Sigma-Aldrich®
Palladium (II) chloride
Formula: PdCl₂
MW: 177.33 g/mol
Melting Pt: 678
Density: 4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003558
CAS Number: 7647-10-1
EINECS: 231-596-2
UN: 3260
ADR: 8 III
Merck Index: 13,07058