Cobalt(II) oxide, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
343153-100G 343153-10G
SIAL343153-100GEA 684 EUR
SIAL343153-100G SIAL343153-10G
Cobalt(II) oxide, Sigma-Aldrich®
Cobalt (II) oxide
Formula: CoO
Melting Pt: 1933 °C
Density: 6.1 g/cm³ (20 °C)
MDL Number: MFCD00016031
CAS Number: 1307-96-6
EINECS: 215-154-6
UN: 2811
ADR: 6.1,III
Merck Index: 12,02507