Cobalt(II) oxide, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Cobalt oxide
M-Clarity™ quality level = MQ100
529443-25G 529443-5G
SIAL529443-25GEA 832 EUR
SIAL529443-25G SIAL529443-5G
Cobalt(II) oxide, Sigma-Aldrich®
Cobalt (II) oxide
Formula: CoO
MW: 74.93 g/mol
Melting Pt: 1933 °C
Density: 6.1 g/cm³ (20 °C)
MDL Number: MFCD00016031
CAS Number: 1307-96-6
EINECS: 215-154-6
UN: 2811
ADR: 6.1,III
Merck Index: 12,02507