Cobalt(II,III) oxide, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Tricobalt tetraoxide, Cobaltic tetraoxide

M-Clarity™ quality level = MQ100

221643-250G 221643-50G
SIAL221643-250GEA 283 EUR
SIAL221643-250G SIAL221643-50G
Cobalt(II,III) oxide, Sigma-Aldrich®
Cobalt (II,III) oxide
Formula: Co₃O₄
Melting Pt: 895 °C
Density: 6.005 g/cm³ (20 °C)
MDL Number: MFCD00010939
CAS Number: 1308-06-1
EINECS: 215-157-2
Merck Index: 13,02454

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR