Iron(III) oxide

Supplier: Roth Carl

ROTH4463.1EA 39.1 EUR
ROTH4463.1
Iron(III) oxide
Iron(III) oxide
Formula: Fe₂O₃
MW: 159,69 g/mol
Melting Pt: 1538 °C
Density: 5,24 g/cm³ (20 °C)
MDL Number: MFCD00011008
CAS Number: 1309-37-1
Merck Index: 12,04072

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR