Nickel (II) oxide, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
481793-25G 481793-5G
SIAL481793-25GEA 245 EUR
SIAL481793-25G SIAL481793-5G
Nickel (II) oxide, Sigma-Aldrich®
Nickel (II) oxide
Formula: NiO
Melting Pt: 1990 °C
Density: 4.6…6.67 g/cm³ (20 °C)
MDL Number: MFCD00011145
CAS Number: 1313-99-1
EINECS: 215-215-7