Nickel (II) oxide, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
637130-100G 637130-25G
SIAL637130-100GEA 245 EUR
SIAL637130-100G SIAL637130-25G
Nickel (II) oxide, Sigma-Aldrich®
Nickel (II) oxide
Formula: NiO
Melting Pt: 1990 °C
Density: 4.6…6.67 g/cm³ (20 °C)
MDL Number: MFCD00011145
CAS Number: 1313-99-1
EINECS: 215-215-7